missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(-)-1-Benzyl-3-aminopyrrolidine 98.0+%, TCI America™

Catalog No. p-7117285
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A117310G 10 g
A117325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A117310G Supplier TCI America Supplier No. A117310G

Chemical Identifiers

CAS 114715-39-8
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00082638
InChI Key HBVNLKQGRZPGRP-LLVKDONJSA-N
Synonym r---1-benzyl-3-aminopyrrolidine, r-1-benzyl-3-aminopyrrolidine, r-1-benzylpyrrolidin-3-amine, 3r---1-benzyl-3-aminopyrrolidine, 3r-1-benzylpyrrolidin-3-amine, r-3-amino-1-n-benzyl-pyrrolidine, r-n-benzyl-3-aminopyrrolidine, 3-pyrrolidinamine, 1-phenylmethyl-, 3r, r-3-amino-1-benzylpyrrolidine, 3r-1-benzyl-3-pyrrolidinamine
PubChem CID 1519354
IUPAC Name (3R)-1-benzylpyrrolidin-3-amine
SMILES N[C@@H]1CCN(CC2=CC=CC=C2)C1

Specifications

Color Yellow
Boiling Point 279°C
Quantity 10 g
UN Number 2735
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3R)-(-)-1-Benzyl-3-aminopyrrolidine