Learn More
3-Vinylbenzoic acid, 96%
CAS: 28447-20-3 | C9H8O2 | 148.161 g/mol
$123.58 - $370.71
Chemical Identifiers
| CAS | 28447-20-3 |
|---|---|
| Molecular Formula | C9H8O2 |
| Molecular Weight (g/mol) | 148.161 |
| MDL Number | MFCD00157038 |
| InChI Key | VWXZFDWVWMQRQR-UHFFFAOYSA-N |
| PubChem CID | 4438231 |
| IUPAC Name | 3-ethenylbenzoic acid |
| SMILES | C=CC1=CC(=CC=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3255603
|
Thermo Scientific Chemicals
H3255603 |
1 g |
Each for $123.58
|
|
|||||
|
AAH3255606
|
Thermo Scientific Chemicals
H3255606 |
5 g |
Each for $370.71
|
|
|||||
Description
Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPolymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, ultraviolet radiations, free radical initiators.
Chemical Identifiers
| 28447-20-3 | |
| 148.161 | |
| VWXZFDWVWMQRQR-UHFFFAOYSA-N | |
| 3-ethenylbenzoic acid |
| C9H8O2 | |
| MFCD00157038 | |
| 4438231 | |
| C=CC1=CC(=CC=C1)C(=O)O |
Specifications
| 28447-20-3 | |
| C9H8O2 | |
| 1 g | |
| VWXZFDWVWMQRQR-UHFFFAOYSA-N | |
| 3-ethenylbenzoic acid | |
| 4438231 | |
| 96% |
| 95°C | |
| MFCD00157038 | |
| 3-vinylbenzoic acid, 3-vinyl benzoic acid, 3-ethenyl-benzoic acid, benzoic acid,3-ethenyl, benzoic acid, 3-ethenyl, benzoic acid, 3-ethenyl-9ci | |
| C=CC1=CC(=CC=C1)C(=O)O | |
| 148.161 | |
| 148.16 | |
| 3-Vinylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only