accessibility menu, dialog, popup

UserName

CAS RN 28447-20-3

IUPAC Name: 3-ethenylbenzoic acid
Synonyms: 3-ethenylbenzoic acid
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: VWXZFDWVWMQRQR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(C=C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results