missing translation for 'onlineSavingsMsg'
Learn More

3-Vinylbenzoic acid, 96%

Catalog No. AAH3255606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3255606 5 g
AAH3255603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3255606 Supplier Thermo Scientific Chemicals Supplier No. H3255606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28447-20-3 | C9H8O2 | 148.161 g/mol

Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymers were created by the imprinting of tri-O-acetyladenosine in acrylic polymer matrix with the use of methacrylic acid (MAA) or 3-vinylbenzoic acid (VBA) as a functional monomer and ethylene glycol dimethacrylate as a cross-linking agent. Nine active ester monomers based on 4-vinylbenzoic acid have been synthesized. Under free radical polymerization conditions these monomers could successfully be polymerized yielding reactive polymers

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, ultraviolet radiations, free radical initiators.

Chemical Identifiers

CAS 28447-20-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00157038
InChI Key VWXZFDWVWMQRQR-UHFFFAOYSA-N
PubChem CID 4438231
IUPAC Name 3-ethenylbenzoic acid
SMILES C=CC1=CC(=CC=C1)C(=O)O

Specifications

Melting Point 95°C
Quantity 5 g
Synonym 3-vinylbenzoic acid, 3-vinyl benzoic acid, 3-ethenyl-benzoic acid, benzoic acid,3-ethenyl, benzoic acid, 3-ethenyl, benzoic acid, 3-ethenyl-9ci
Formula Weight 148.16
Percent Purity 96%
Chemical Name or Material 3-Vinylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only