missing translation for 'onlineSavingsMsg'
Learn More

3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one, 97%, Thermo Scientific™

Chemical Identifiers

CAS 34932-07-5
Molecular Formula C22H19O2P
Molecular Weight (g/mol) 346.37
InChI Key JUKRJGQYUNADDD-UHFFFAOYSA-N
Synonym 2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
PubChem CID 97111
IUPAC Name 3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
SMILES C1COC(=O)C1=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC440990010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

34932-07-5
346.37
2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
C22H19O2P
JUKRJGQYUNADDD-UHFFFAOYSA-N
97111
C1COC(=O)C1=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

34932-07-5
100.0
Authentic
Glass bottle
2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
C1COC(=O)C1=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
346.37
346.37
3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one
96.0
73.24 to 79.34% (C)
97%
C22H19O2P
JUKRJGQYUNADDD-UHFFFAOYSA-N
3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
97111
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only