missing translation for 'onlineSavingsMsg'
Learn More

3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 440990010

Quantity 1g

 View more versions of this product

Catalog No. AC440990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

34932-07-5
346.37
2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
C22H19O2P
JUKRJGQYUNADDD-UHFFFAOYSA-N
97111
C1COC(=O)C1=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

3-(Triphenylphosphoranylidene)dihydrofuran-2-(3H)-one
96.0
73.24 to 79.34% (C)
97%
C22H19O2P
2 3h-furanone, 3-triphenylphosphoranylidene, gamma-butyrolactonylidene triphenyl phosphorane, 2-triphenylphosphoranylidene-gamma-butyrolactone, alpha-triphenylphosphoranylidene-gamma-butyrolactone, 2 3h-furanone, dihydro-3-triphenylphosphoranylidene, 3-triphenylphosphoranylidene dihydro-2 3h-furanone, 3-triphenyl-??-phosphanylidene oxolan-2-one, 3-triphenyl-, e5-phosphanylidene dihydrofuran-2 3h-one, .gamma.-butyrolactonylidene triphenyl phosphorane, 2-oxotetrahydrofuran-3-ylidenetriphenylphosphorane
C1COC(=O)C1=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
346.37
346.37
34932-07-5
100.0
Authentic
Glass bottle
1g
JUKRJGQYUNADDD-UHFFFAOYSA-N
3-(triphenyl-$l^{5}-phosphanylidene)oxolan-2-one
97111
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...