accessibility menu, dialog, popup

UserName

OOP

CAS RN 34932-07-5

IUPAC Name: 3-(triphenyl-λ⁵-phosphanylidene)oxolan-2-one
Synonyms: 3-(triphenyl-λ⁵-phosphanylidene)oxolan-2-one
Molecular Weight (g/mol): 346.37
Molecular Formula: C22H19O2P
InChi Key: JUKRJGQYUNADDD-UHFFFAOYSA-N
SMILES: O=C1OCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)