missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Trimethoxysilylpropyl Chloride 97.0+%, TCI America™
Supplier: TCI America C0840100ML
Specifications
| 3-Trimethoxysilylpropyl Chloride | |
| -50°C | |
| 195°C | |
| MFCD00000997 | |
| 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
| CO[Si](CCCCl)(OC)OC | |
| 198.718 | |
| 198.72 | |
| Liquid |
| 2530-87-2 | |
| Yellow | |
| C6H15ClO3Si | |
| 100 mL | |
| OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
| 3-chloropropyl(trimethoxy)silane | |
| 62449 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 2530-87-2 | |
| 198.718 | |
| OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
| 62449 | |
| CO[Si](CCCCl)(OC)OC |
| C6H15ClO3Si | |
| MFCD00000997 | |
| 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
| 3-chloropropyl(trimethoxy)silane |
Safety and Handling
EINECSNumber : (2)-2079
RTECSNumber : VV2680000
TSCA : Yes