missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethoxysilylpropyl Chloride 97.0+%, TCI America™

Catalog No. p-7124638
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C084025ML 25 mL
C0840100ML 100 mL
C0840500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C084025ML Supplier TCI America Supplier No. C084025ML
Only null left

Chemical Identifiers

CAS 2530-87-2
Molecular Formula C6H15ClO3Si
Molecular Weight (g/mol) 198.718
MDL Number MFCD00000997
InChI Key OXYZDRAJMHGSMW-UHFFFAOYSA-N
Synonym 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
PubChem CID 62449
IUPAC Name 3-chloropropyl(trimethoxy)silane
SMILES CO[Si](CCCCl)(OC)OC

Specifications

Melting Point -50°C
Color Yellow
Boiling Point 195°C
Quantity 25 mL
Formula Weight 198.72
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Trimethoxysilylpropyl Chloride
EINECSNumber (2)-2079
RTECSNumber VV2680000
TSCA Yes