accessibility menu, dialog, popup

UserName

CAS RN 2530-87-2

IUPAC Name: (3-chloropropyl)trimethoxysilane
Synonyms: (3-chloropropyl)trimethoxysilane
Molecular Weight (g/mol): 198.72
Molecular Formula: C6H15ClO3Si
InChi Key: OXYZDRAJMHGSMW-UHFFFAOYSA-N
SMILES: CO[Si](CCCCl)(OC)OC
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (8)
  • (8)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (3)
  • (3)