Learn More
3-(Trifluoromethylthio)aniline, 96%
CAS: 369-68-6 | C7H6F3NS | 193.19 g/mol
$156.23 - $616.10
Chemical Identifiers
| CAS | 369-68-6 |
|---|---|
| Molecular Formula | C7H6F3NS |
| Molecular Weight (g/mol) | 193.19 |
| MDL Number | MFCD00190130 |
| InChI Key | DENPAKQJZNDKEL-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethylthio aniline, 3-trifluoromethyl sulfanyl aniline, 3-aminophenyl trifluoromethyl sulphide, 3-aminophenyl trifluoromethyl sulfide, 3-trifluoromethyl thio aniline, 3-trifluoromethylsulfanyl aniline, benzenamine, 3-trifluoromethyl thio, benzeneamine, 3-trifluoromethyl thio, 3-trifluoromethylthio phenylamine |
| PubChem CID | 2735939 |
| IUPAC Name | 3-(trifluoromethylsulfanyl)aniline |
| SMILES | NC1=CC=CC(SC(F)(F)F)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2373403
|
Thermo Scientific Chemicals
B2373403 |
1 g |
Each for $156.23
|
|
|||||
|
AAB2373406
|
Thermo Scientific Chemicals
B2373406 |
5 g |
Each for $616.10
|
|
|||||
Description
3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
| 369-68-6 | |
| 193.19 | |
| DENPAKQJZNDKEL-UHFFFAOYSA-N | |
| 2735939 | |
| NC1=CC=CC(SC(F)(F)F)=C1 |
| C7H6F3NS | |
| MFCD00190130 | |
| 3-trifluoromethylthio aniline, 3-trifluoromethyl sulfanyl aniline, 3-aminophenyl trifluoromethyl sulphide, 3-aminophenyl trifluoromethyl sulfide, 3-trifluoromethyl thio aniline, 3-trifluoromethylsulfanyl aniline, benzenamine, 3-trifluoromethyl thio, benzeneamine, 3-trifluoromethyl thio, 3-trifluoromethylthio phenylamine | |
| 3-(trifluoromethylsulfanyl)aniline |
Specifications
| 369-68-6 | |
| 221°C to 224°C | |
| Unpleasant | |
| 1.522 | |
| 1 g | |
| 3-trifluoromethylthio aniline, 3-trifluoromethyl sulfanyl aniline, 3-aminophenyl trifluoromethyl sulphide, 3-aminophenyl trifluoromethyl sulfide, 3-trifluoromethyl thio aniline, 3-trifluoromethylsulfanyl aniline, benzenamine, 3-trifluoromethyl thio, benzeneamine, 3-trifluoromethyl thio, 3-trifluoromethylthio phenylamine | |
| DENPAKQJZNDKEL-UHFFFAOYSA-N | |
| 3-(trifluoromethylsulfanyl)aniline | |
| 2735939 | |
| 96% |
| 1.348 | |
| 103°C (218°F) | |
| C7H6F3NS | |
| MFCD00190130 | |
| UN2810 | |
| Immiscible with water. | |
| NC1=CC=CC(SC(F)(F)F)=C1 | |
| 193.19 | |
| 193.19 | |
| 3-(Trifluoromethylthio)aniline |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 206-722-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only