accessibility menu, dialog, popup

UserName

NH2SFFF

CAS RN 369-68-6

IUPAC Name: 3-[(trifluoromethyl)sulfanyl]aniline
Synonyms: 3-[(trifluoromethyl)sulfanyl]aniline
Molecular Weight (g/mol): 193.19
Molecular Formula: C7H6F3NS
InChi Key: DENPAKQJZNDKEL-UHFFFAOYSA-N
SMILES: NC1=CC=CC(SC(F)(F)F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)