missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethylthio)aniline, 96%

Catalog No. AAB2373403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2373403 1 g
AAB2373406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2373403 Supplier Thermo Scientific Chemicals Supplier No. B2373403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 369-68-6 | C7H6F3NS | 193.19 g/mol

3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 369-68-6
Molecular Formula C7H6F3NS
Molecular Weight (g/mol) 193.19
MDL Number MFCD00190130
InChI Key DENPAKQJZNDKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethylthio aniline, 3-trifluoromethyl sulfanyl aniline, 3-aminophenyl trifluoromethyl sulphide, 3-aminophenyl trifluoromethyl sulfide, 3-trifluoromethyl thio aniline, 3-trifluoromethylsulfanyl aniline, benzenamine, 3-trifluoromethyl thio, benzeneamine, 3-trifluoromethyl thio, 3-trifluoromethylthio phenylamine
PubChem CID 2735939
IUPAC Name 3-(trifluoromethylsulfanyl)aniline
SMILES NC1=CC=CC(SC(F)(F)F)=C1

Specifications

Density 1.348
Boiling Point 221°C to 224°C
Flash Point 103°C (218°F)
Odor Unpleasant
Refractive Index 1.522
Quantity 1 g
UN Number UN2810
Solubility Information Immiscible with water.
Formula Weight 193.19
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethylthio)aniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-722-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only