missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethylbenzoic anhydride, 97%

Catalog No. AAH3377906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3377906 5 g
AAH3377903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3377906 Supplier Thermo Scientific Chemicals Supplier No. H3377906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25753-15-5 | C16H8F6O3 | 362.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25753-15-5
Molecular Formula C16H8F6O3
Molecular Weight (g/mol) 362.23
MDL Number MFCD09757594
InChI Key BYLMYFXAIMTZIC-UHFFFAOYSA-N
Synonym 3-trifluoromethylbenzoic anhydride, 3-trifluoromethyl benzoic anhydride, benzoic acid, 3-trifluoromethyl-, anhydride, 3-trifluoromethyl benzoyl 3-trifluoromethyl benzoate, acmc-1cd8h, 3-trifluoromethyl phenyl anhydride #, benzoic acid,3-trifluoromethyl-, anhydride with 3-trifluoromethyl benzoic acid
PubChem CID 599431
IUPAC Name [3-(trifluoromethyl)benzoyl] 3-(trifluoromethyl)benzoate
SMILES FC(F)(F)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 362.23
Percent Purity 97%
Chemical Name or Material 3-Trifluoromethylbenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only