CAS RN 25753-15-5
IUPAC Name:
3-(trifluoromethyl)benzoyl 3-(trifluoromethyl)benzoate
Synonyms:
3-(trifluoromethyl)benzoyl 3-(trifluoromethyl)benzoate
Molecular Weight (g/mol):
362.23
Molecular Formula:
C16H8F6O3
InChi Key:
BYLMYFXAIMTZIC-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Trifluoromethylbenzoic anhydride, 97%
CAS: 25753-15-5 Molecular Formula: C16H8F6O3 Molecular Weight (g/mol): 362.23 MDL Number: MFCD09757594 InChI Key: BYLMYFXAIMTZIC-UHFFFAOYSA-N Synonym: 3-trifluoromethylbenzoic anhydride,3-trifluoromethyl benzoic anhydride,benzoic acid, 3-trifluoromethyl-, anhydride,3-trifluoromethyl benzoyl 3-trifluoromethyl benzoate,acmc-1cd8h,3-trifluoromethyl phenyl anhydride #,benzoic acid,3-trifluoromethyl-, anhydride with 3-trifluoromethyl benzoic acid PubChem CID: 599431 IUPAC Name: [3-(trifluoromethyl)benzoyl] 3-(trifluoromethyl)benzoate SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)C(F)(F)F