missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)thiophenol, 97%

Catalog No. AAL0719103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0719103 1 g
AAL0719106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0719103 Supplier Thermo Scientific Chemicals Supplier No. L0719103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 937-00-8 | C7H5F3S | 178.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 937-00-8
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.17
MDL Number MFCD00041142
InChI Key SCURCOWZQJIUGR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl thiophenol, 3-trifluoromethyl benzenethiol, 3-trifluoromethyl benzene-1-thiol, 3-trifluoromethylthiophenol, m-trifluoromethylthiophenol, 3-mercaptobenzotrifluoride, m-trifluoromethyl benzenethiol, benzenethiol, 3-trifluoromethyl, pubchem4600
PubChem CID 136751
IUPAC Name 3-(trifluoromethyl)benzenethiol
SMILES FC(F)(F)C1=CC=CC(S)=C1

Specifications

Boiling Point 71°C to 72°C (20 mmHg)
Odor Stench
Refractive Index 1.49
Quantity 1 g
UN Number UN2810
Beilstein 1943114
Sensitivity Air Sensitive
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)thiophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only