missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyltrimethylammonium Iodide 98.0+%, TCI America™

Catalog No. T101425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T101425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T101425G Supplier TCI America Supplier No. T101425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27389-57-7
Molecular Formula C10H13F3IN
Molecular Weight (g/mol) 331.121
MDL Number MFCD00059980
InChI Key VKZIUXSJJSEBAK-UHFFFAOYSA-M
Synonym m-TFPTAI, Trimethyl-3-trifluoromethylphenylammonium Iodide
PubChem CID 2760773
IUPAC Name trimethyl-[3-(trifluoromethyl)phenyl]azanium;iodide
SMILES C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[I-]

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 25 g
Formula Weight 331.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)phenyltrimethylammonium Iodide