missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyltrimethylammonium hydroxide, 5% w/v in methanol

Catalog No. AA43284AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43284AA 5 mL
AA43284AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43284AA Supplier Thermo Scientific Chemicals Supplier No. 043284AA

Transesterification reactions | CAS: 68254-41-1 | C10H14F3NO | 221.223 g/mol

Transesterification reactions.3-(Trifluoromethyl)phenyltrimethylammonium hydroxide is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68254-41-1
Molecular Formula C10H14F3NO
Molecular Weight (g/mol) 221.223
MDL Number MFCD00059473
InChI Key BFPOZPZYPNVMHU-UHFFFAOYSA-M
Synonym 3-trifluoromethyl phenyltrimethylammonium hydroxide, m-tfptah, trifluoromethylphenyltrimethylammoniumhydroxide, 3-trifluoromethyl phenyltrimethylammonium hydroxide in methanol, n,n,n-trimethyl-3-trifluoromethyl anilinium hydroxide, trimethyl-3-trifluoromethyl phenyl azanium hydroxide, m-trifluoromethylphenyl trimethylammonium hydroxide, trimethyl-3-trifluoromethylphenylammonium hydroxide, n,n,n-trimethyl-3-trifluoromethyl anilinium oxidanide, trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium hydroxide
PubChem CID 2760772
IUPAC Name trimethyl-[3-(trifluoromethyl)phenyl]azanium;hydroxide
SMILES C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[OH-]

Specifications

Quantity 5 mL
UN Number UN2924
Solubility Information Miscible with water and alcohols.
Formula Weight 221.22
Concentration or Composition (by Analyte or Components) 5% w/v in methanol
Physical Form Liquid
Chemical Name or Material 3- (Trifluoromethyl)phenyltrimethylammonium hydroxide
Hazard Category H225-H301+H311+H331-H370
Hazard Statement GHS H Statement
H224-H300-H310-H330-H370-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 200-659-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only