missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyltrimethylammonium Bromide 98.0+%, TCI America™

Catalog No. T103625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T103625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T103625G Supplier TCI America Supplier No. T103625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 262608-95-7
Molecular Formula C10H13BrF3N
Molecular Weight (g/mol) 284.12
MDL Number MFCD00059979
InChI Key KPFRXMSETZXGKJ-UHFFFAOYSA-M
Synonym Trimethyl-3-trifluoromethylphenylammonium Bromide
PubChem CID 2760770
IUPAC Name trimethyl-[3-(trifluoromethyl)phenyl]azanium;bromide
SMILES C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 284.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)phenyltrimethylammonium Bromide