accessibility menu, dialog, popup

UserName

CH3NH3CCH3FFFBr

CAS RN 262608-95-7

IUPAC Name: N,N,N-trimethyl-3-(trifluoromethyl)anilinium bromide
Synonyms: N,N,N-trimethyl-3-(trifluoromethyl)anilinium bromide
Molecular Weight (g/mol): 284.12
Molecular Formula: C10H13BrF3N
InChi Key: KPFRXMSETZXGKJ-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results