missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylhydrazine 97.0+%, TCI America™

Catalog No. T24111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24111G 1 g
T24115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24111G Supplier TCI America Supplier No. T24111G

Chemical Identifiers

CAS 368-78-5
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00025093
InChI Key RSESUCWJKLHXEZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylhydrazine, 3-trifluoromethyl phenyl hydrazine, 3-trifluoromethylphenylhydrazine, hydrazine, 3-trifluoromethyl phenyl, 3-trifloromethylphenylhydrazine, 3-trifluouromethyl phenylhydrazinehydrochloride, 3-trifluoromethyl phenylhydrazine hydrochioride, pubchem3280, enamine_001261
PubChem CID 98650
IUPAC Name [3-(trifluoromethyl)phenyl]hydrazine
SMILES C1=CC(=CC(=C1)NN)C(F)(F)F

Specifications

Color Yellow
Boiling Point 83°C
Quantity 1 g
Formula Weight 176.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine