accessibility menu, dialog, popup

UserName

NH2HNFFF

CAS RN 368-78-5

IUPAC Name: [3-(trifluoromethyl)phenyl]hydrazine
Synonyms: [3-(trifluoromethyl)phenyl]hydrazine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: RSESUCWJKLHXEZ-UHFFFAOYSA-N
SMILES: NNC1=CC=CC(=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)