missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylhydrazine, 95%

Catalog No. AAL1199606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1199606 5 g
AAL1199603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1199606 Supplier Thermo Scientific Chemicals Supplier No. L1199606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 368-78-5 | C7H7F3N2 | 176.142 g/mol

3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 368-78-5
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00025093
InChI Key RSESUCWJKLHXEZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylhydrazine, 3-trifluoromethyl phenyl hydrazine, 3-trifluoromethylphenylhydrazine, hydrazine, 3-trifluoromethyl phenyl, 3-trifloromethylphenylhydrazine, 3-trifluouromethyl phenylhydrazinehydrochloride, 3-trifluoromethyl phenylhydrazine hydrochioride, pubchem3280, enamine_001261
PubChem CID 98650
IUPAC Name [3-(trifluoromethyl)phenyl]hydrazine
SMILES C1=CC(=CC(=C1)NN)C(F)(F)F

Specifications

Density 1.348
Boiling Point 80°C to 83°C (9 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.504
Quantity 5 g
Beilstein 640075
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 176.14
Percent Purity 95%
Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-713-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only