missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl bromide, 98%

Catalog No. aaa1933014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1933014 25 g
AAA1933006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1933014 Supplier Thermo Scientific Chemicals Supplier No. A1933014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 402-23-3 | C8H6BrF3 | 239.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-23-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.04
MDL Number MFCD00000395
InChI Key MYYYZNVAUZVXBO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl bromide, 1-bromomethyl-3-trifluoromethyl benzene, m-trifluoromethylbenzyl bromide, 3-trifluoromethylbenzyl bromide, 3-bromomethyl benzotrifluoride, 3-trifluoromethylbenzylbromide, 3-trifluoromethyl benzylbromide, 1-bromomethyl-3-trifluoromethyl-benzene, alpha'-bromo-alpha,alpha,alpha-trifluoro-m-xylene
PubChem CID 123061
IUPAC Name 1-(bromomethyl)-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(CBr)=C1

Specifications

Density 1.565
Boiling Point 67°C to 69°C (4 mmHg)
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.492
Quantity 25 g
UN Number UN3265
Beilstein 743243
Formula Weight 239.04
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzyl bromide
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 206-938-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only