accessibility menu, dialog, popup

UserName

CAS RN 402-23-3

IUPAC Name: 1-(bromomethyl)-3-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: MYYYZNVAUZVXBO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CBr)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)