missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™

Supplier:  TCI America T133525G

 View more versions of this product

Catalog No. T133525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-(Trifluoromethyl)benzoyl Chloride
Yellow
C8H4ClF3O
25 g
3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl
208.564
208.56
Liquid
2251-65-2
85°C
MFCD00000680
3265
RUJYJCANMOTJMO-UHFFFAOYSA-N
3-(trifluoromethyl)benzoyl chloride
75257
≥98.0% (GC,T)

Chemical Identifiers

2251-65-2
208.564
RUJYJCANMOTJMO-UHFFFAOYSA-N
75257
C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl
C8H4ClF3O
MFCD00000680
3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
3-(trifluoromethyl)benzoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions