missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7136719
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13355G 5 g
T133525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13355G Supplier TCI America Supplier No. T13355G
Only null left

Chemical Identifiers

CAS 2251-65-2
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00000680
InChI Key RUJYJCANMOTJMO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
PubChem CID 75257
IUPAC Name 3-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 3265
Formula Weight 208.56
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzoyl Chloride