missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. C100325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C100325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C100325G Supplier TCI America Supplier No. C100325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 368-77-4
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001807
InChI Key OGOBINRVCUWLGN-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzonitrile, 3-cyanobenzotrifluoride, m-trifluoromethylbenzonitrile, alpha,alpha,alpha-trifluoro-m-tolunitrile, benzonitrile, 3-trifluoromethyl, a,a,a-trifluoro-m-tolunitrile, unii-y7zd7dk3ay, alpha,alpha,alpha-trifluoro-m-toluonitrile, y7zd7dk3ay, a,a,a-trifluor-m-tolunitril
PubChem CID 67783
IUPAC Name 3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=CC(=C1)C(F)(F)F)C#N

Specifications

Color Colorless
Boiling Point 189°C
Quantity 25 g
UN Number 3276
Formula Weight 171.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzonitrile