missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzenesulfonyl chloride, 95%, Thermo Scientific™

Catalog No. AC306280010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC306280010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC306280010 Supplier Thermo Scientific Chemicals Supplier No. 306280010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 777-44-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.62
MDL Number MFCD00014724
InChI Key ONCAZCNPWWQQMW-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
PubChem CID 2733250
IUPAC Name 3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.5200g/mL
Boiling Point 77°C to 79°C (0.8 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4SO2Cl
Refractive Index 1.4847 to 1.4867
Quantity 1 g
Specific Gravity 1.52
Solubility Information Solubility in water: reacts
Formula Weight 244.62
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonyl chloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
TSCA TSCA

RUO – Research Use Only