accessibility menu, dialog, popup

UserName

CAS RN 777-44-6

IUPAC Name: 3-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 244.61
Molecular Formula: C7H4ClF3O2S
InChi Key: ONCAZCNPWWQQMW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)