Learn More
3-(Trifluoromethyl)aniline, 99%
CAS: 98-16-8 | C7H6F3N | 161.127 g/mol
Supplier: Thermo Scientific Chemicals A1591036
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediate.
Solubility
Soluble in water. [5 g/L (20°C)]
Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications
| 3-(Trifluoromethyl)aniline | |
| 5°C to 6°C | |
| 187°C to 189°C | |
| Recognizable | |
| 1.48 | |
| 500 g | |
| 387672 | |
| Soluble in water. [5g/L (20°C)] | |
| C1=CC(=CC(=C1)N)C(F)(F)F | |
| 161.127 | |
| 161.13 |
| 98-16-8 | |
| 1.295 | |
| 85°C (185°F) | |
| C7H6F3N | |
| MFCD00007797 | |
| UN2948 | |
| 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline | |
| VIUDTWATMPPKEL-UHFFFAOYSA-N | |
| 3-(trifluoromethyl)aniline | |
| 7375 | |
| 99% |
Chemical Identifiers
| 98-16-8 | |
| 161.127 | |
| VIUDTWATMPPKEL-UHFFFAOYSA-N | |
| 7375 | |
| C1=CC(=CC(=C1)N)C(F)(F)F |
| C7H6F3N | |
| MFCD00007797 | |
| 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline | |
| 3-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H330-H373-H227-H302-H312-H319
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312-H315-H318-H330-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-TRIFLUOROMETHYLANILINE
EINECSNumber : 202-643-4
RTECSNumber : XU9180000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only