missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)aniline, 99%

Catalog No. p-7022923
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1591022 100 g
AAA1591036 500 g
AAA159100E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1591022 Supplier Thermo Scientific Chemicals Supplier No. A1591022

missing translation for 'validMainframeMsg'

CAS: 98-16-8 | C7H6F3N | 161.127 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. [5 g/L (20°C)]

Notes
Store in cool dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 98-16-8
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007797
InChI Key VIUDTWATMPPKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
PubChem CID 7375
IUPAC Name 3-(trifluoromethyl)aniline
SMILES C1=CC(=CC(=C1)N)C(F)(F)F

Specifications

Melting Point 5°C to 6°C
Density 1.295
Boiling Point 187°C to 189°C
Flash Point 85°C (185°F)
Odor Recognizable
Refractive Index 1.48
Quantity 100 g
UN Number UN2948
Beilstein 387672
Solubility Information Soluble in water. [5g/L (20°C)]
Formula Weight 161.13
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)aniline
Show More Show Less
Hazard Category H227-H302+H312-H315-H318-H330-H335-H373
Hazard Statement GHS H Statement
H330-H373-H227-H302-H312-H319
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-TRIFLUOROMETHYLANILINE
EINECSNumber 202-643-4
RTECSNumber XU9180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only