missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethoxy)benzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America T19555G
Specifications
| 3-(Trifluoromethoxy)benzyl Alcohol | |
| Yellow | |
| MFCD00061270 | |
| 3-trifluoromethoxy benzyl alcohol, 3-trifluoromethoxy phenyl methanol, 3-trifluoromethoxybenzyl alcohol, m-trifluoromethoxybenzyl alcohol, benzenemethanol, 3-trifluoromethoxy, 3-trifluoromethoxy phenyl methan-1-ol, pubchem4682, acmc-1an32, ksc494a4t | |
| OCC1=CC=CC(OC(F)(F)F)=C1 | |
| 192.14 | |
| 192.14 | |
| Liquid |
| 50823-90-0 | |
| C8H7F3O2 | |
| 5 g | |
| XRGSSROOYSFMMS-UHFFFAOYSA-N | |
| [3-(trifluoromethoxy)phenyl]methanol | |
| 142783 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 50823-90-0 | |
| 192.14 | |
| XRGSSROOYSFMMS-UHFFFAOYSA-N | |
| 142783 | |
| OCC1=CC=CC(OC(F)(F)F)=C1 |
| C8H7F3O2 | |
| MFCD00061270 | |
| 3-trifluoromethoxy benzyl alcohol, 3-trifluoromethoxy phenyl methanol, 3-trifluoromethoxybenzyl alcohol, m-trifluoromethoxybenzyl alcohol, benzenemethanol, 3-trifluoromethoxy, 3-trifluoromethoxy phenyl methan-1-ol, pubchem4682, acmc-1an32, ksc494a4t | |
| [3-(trifluoromethoxy)phenyl]methanol |
Safety and Handling
TSCA : No