missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethoxy)benzyl Alcohol 98.0+%, TCI America™
$104.38 - $311.30
Chemical Identifiers
| CAS | 50823-90-0 |
|---|---|
| Molecular Formula | C8H7F3O2 |
| Molecular Weight (g/mol) | 192.14 |
| MDL Number | MFCD00061270 |
| InChI Key | XRGSSROOYSFMMS-UHFFFAOYSA-N |
| Synonym | 3-trifluoromethoxy benzyl alcohol, 3-trifluoromethoxy phenyl methanol, 3-trifluoromethoxybenzyl alcohol, m-trifluoromethoxybenzyl alcohol, benzenemethanol, 3-trifluoromethoxy, 3-trifluoromethoxy phenyl methan-1-ol, pubchem4682, acmc-1an32, ksc494a4t |
| PubChem CID | 142783 |
| IUPAC Name | [3-(trifluoromethoxy)phenyl]methanol |
| SMILES | OCC1=CC=CC(OC(F)(F)F)=C1 |
Chemical Identifiers
| 50823-90-0 | |
| 192.14 | |
| XRGSSROOYSFMMS-UHFFFAOYSA-N | |
| 142783 | |
| OCC1=CC=CC(OC(F)(F)F)=C1 |
| C8H7F3O2 | |
| MFCD00061270 | |
| 3-trifluoromethoxy benzyl alcohol, 3-trifluoromethoxy phenyl methanol, 3-trifluoromethoxybenzyl alcohol, m-trifluoromethoxybenzyl alcohol, benzenemethanol, 3-trifluoromethoxy, 3-trifluoromethoxy phenyl methan-1-ol, pubchem4682, acmc-1an32, ksc494a4t | |
| [3-(trifluoromethoxy)phenyl]methanol |
Specifications
| 50823-90-0 | |
| C8H7F3O2 | |
| 1 g | |
| XRGSSROOYSFMMS-UHFFFAOYSA-N | |
| [3-(trifluoromethoxy)phenyl]methanol | |
| 142783 | |
| ≥98.0% (GC) | |
| 3-(Trifluoromethoxy)benzyl Alcohol |
| Yellow | |
| MFCD00061270 | |
| 3-trifluoromethoxy benzyl alcohol, 3-trifluoromethoxy phenyl methanol, 3-trifluoromethoxybenzyl alcohol, m-trifluoromethoxybenzyl alcohol, benzenemethanol, 3-trifluoromethoxy, 3-trifluoromethoxy phenyl methan-1-ol, pubchem4682, acmc-1an32, ksc494a4t | |
| OCC1=CC=CC(OC(F)(F)F)=C1 | |
| 192.14 | |
| 192.14 | |
| Liquid |
Safety and Handling
TSCA : No