missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Thiopheneethanol 97.0+%, TCI America™
Supplier: TCI America T147525G
Specifications
| 3-Thiopheneethanol | |
| Yellow | |
| C6H8OS | |
| 25 g | |
| YYPNNBPPDFTQFX-UHFFFAOYSA-N | |
| 2-(thiophen-3-yl)ethan-1-ol | |
| 83731 | |
| ≥97.0% (GC) |
| 13781-67-4 | |
| 111°C | |
| MFCD00009766 | |
| 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 | |
| OCCC1=CSC=C1 | |
| 128.19 | |
| 128.19 | |
| Liquid |
Chemical Identifiers
| 13781-67-4 | |
| 128.19 | |
| YYPNNBPPDFTQFX-UHFFFAOYSA-N | |
| 83731 | |
| OCCC1=CSC=C1 |
| C6H8OS | |
| MFCD00009766 | |
| 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 | |
| 2-(thiophen-3-yl)ethan-1-ol |
Safety and Handling
TSCA : No