missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneethanol 97.0+%, TCI America™

Catalog No. p-7136795
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14755G 5 g
T147525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14755G Supplier TCI America Supplier No. T14755G

Chemical Identifiers

CAS 13781-67-4
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00009766
InChI Key YYPNNBPPDFTQFX-UHFFFAOYSA-N
Synonym 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415
PubChem CID 83731
IUPAC Name 2-(thiophen-3-yl)ethan-1-ol
SMILES OCCC1=CSC=C1

Specifications

Color Yellow
Boiling Point 111°C
Quantity 5 g
Formula Weight 128.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Thiopheneethanol