missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylphenol 98.0+%, TCI America™

Catalog No. B073125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B073125G 25 g
B0731500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B073125G Supplier TCI America Supplier No. B073125G

Chemical Identifiers

CAS 585-34-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002300
InChI Key CYEKUDPFXBLGHH-UHFFFAOYSA-N
Synonym m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
PubChem CID 11450
ChEBI CHEBI:34348
IUPAC Name 3-tert-butylphenol
SMILES CC(C)(C)C1=CC=CC(O)=C1

Specifications

Melting Point 41°C
Color White
Boiling Point 132°C
Quantity 25 g
UN Number 2430
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-tert-Butylphenol
EINECSNumber (3)-0503
TSCA Yes