accessibility menu, dialog, popup

UserName

CAS RN 585-34-2

IUPAC Name: 3-tert-butylphenol
Synonyms: 3-tert-butylphenol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: CYEKUDPFXBLGHH-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=CC(O)=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)