missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyldimethylsiloxy)thiophenol, 95%, Thermo Scientific Chemicals

Catalog No. AAL1723803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1723803 1 g
AAL1723806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1723803 Supplier Thermo Scientific Chemicals Supplier No. L1723803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216393-56-5
Molecular Formula C12H20OSSi
Molecular Weight (g/mol) 240.44
MDL Number MFCD01318115
InChI Key KRHCTXFJCFMFOR-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyl oxy benzenethiol, 3-tert-butyldimethylsiloxy thiophenol, acmc-20ani3, 3-tert-butyl-dimethylsiloxy-thiophenol, 3-tert-butyldimethylsilyloxy benzenethiol, 3-t-butyl-dimethyl-silanyloxy-benzenethiol, 3-tert-butyl-dimethyl-silanyloxy-benzenethiol, 3-tert-butyl dimethyl silyl oxy benzene-1-thiol, 3-1,1-dimethylethyl dimethyl silyl oxy benzenethiol
PubChem CID 21976868
SMILES CC(C)(C)[Si](C)(C)OC1=CC(S)=CC=C1

Specifications

Refractive Index 1.521
Quantity 1 g
Sensitivity Air Sensitive
IUPAC Name 3-[(tert-butyldimethylsilyl)oxy]benzene-1-thiol
Formula Weight 240.44
Percent Purity 95%
Chemical Name or Material 3-(tert-Butyldimethylsiloxy)thiophenol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only