missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pyridazinecarboxylic acid, 97%, Thermo Scientific™
$175.89 - $573.47
Chemical Identifiers
| CAS | 2164-61-6 |
|---|---|
| Molecular Formula | C5H4N2O2 |
| Molecular Weight (g/mol) | 124.1 |
| InChI Key | RUUOPSRRIKJHNH-UHFFFAOYSA-N |
| Synonym | 3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid; |
| PubChem CID | 269369 |
| IUPAC Name | pyridazine-3-carboxylic acid |
| SMILES | C1=CC(=NN=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC446720010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC446720050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2164-61-6 | |
| 124.1 | |
| 3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid; | |
| pyridazine-3-carboxylic acid |
| C5H4N2O2 | |
| RUUOPSRRIKJHNH-UHFFFAOYSA-N | |
| 269369 | |
| C1=CC(=NN=C1)C(=O)O |
Specifications
| 2164-61-6 | |
| Glass bottle | |
| 3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid; | |
| C1=CC(=NN=C1)C(=O)O | |
| 124.1 | |
| 124.1 | |
| 3-Pyridazinecarboxylic acid |
| 97% | |
| C5H4N2O2 | |
| RUUOPSRRIKJHNH-UHFFFAOYSA-N | |
| pyridazine-3-carboxylic acid | |
| 269369 | |
| 97% |
Safety and Handling
Warning
RUO – Research Use Only