missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridazinecarboxylic acid, 97%

Catalog No. AC446720050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446720050 5 g
AC446720010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446720050 Supplier Thermo Scientific Chemicals Supplier No. 446720050

CAS: 2164-61-6 | C5H4N2O2 | 124.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2164-61-6
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.1
InChI Key RUUOPSRRIKJHNH-UHFFFAOYSA-N
Synonym 3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid;
PubChem CID 269369
IUPAC Name pyridazine-3-carboxylic acid
SMILES C1=CC(=NN=C1)C(=O)O

Specifications

Melting Point 190.8°C to 194.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material 3-Pyridazinecarboxylic acid
Hazard Category Warning

RUO – Research Use Only