accessibility menu, dialog, popup

UserName

HOONN

CAS RN 2164-61-6

IUPAC Name: pyridazine-3-carboxylic acid
Synonyms: pyridazine-3-carboxylic acid
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: RUUOPSRRIKJHNH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results