missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylsalicylic Acid 98.0+%, TCI America™
$165.90 - $475.33
Chemical Identifiers
| CAS | 304-06-3 |
|---|---|
| Molecular Formula | C13H10O3 |
| Molecular Weight (g/mol) | 214.22 |
| MDL Number | MFCD00020257 |
| InChI Key | ZJWUEJOPKFYFQD-UHFFFAOYSA-N |
| Synonym | 2-Hydroxybiphenyl-3-carboxylic Acid |
| PubChem CID | 9348 |
| IUPAC Name | 2-hydroxy-3-phenylbenzoic acid |
| SMILES | C1=CC=C(C=C1)C2=C(C(=CC=C2)C(=O)O)O |
Chemical Identifiers
| 304-06-3 | |
| 214.22 | |
| ZJWUEJOPKFYFQD-UHFFFAOYSA-N | |
| 9348 | |
| C1=CC=C(C=C1)C2=C(C(=CC=C2)C(=O)O)O |
| C13H10O3 | |
| MFCD00020257 | |
| 2-Hydroxybiphenyl-3-carboxylic Acid | |
| 2-hydroxy-3-phenylbenzoic acid |
Specifications
| 304-06-3 | |
| White-Yellow | |
| MFCD00020257 | |
| 2-Hydroxybiphenyl-3-carboxylic Acid | |
| C1=CC=C(C=C1)C2=C(C(=CC=C2)C(=O)O)O | |
| 214.22 | |
| 214.22 | |
| Crystalline Powder |
| 191°C | |
| C13H10O3 | |
| 5 g | |
| ZJWUEJOPKFYFQD-UHFFFAOYSA-N | |
| 2-hydroxy-3-phenylbenzoic acid | |
| 9348 | |
| ≥98.0% (T) | |
| 3-Phenylsalicylic Acid |
Safety and Handling
EINECSNumber : (4)-0023
RTECSNumber : VO5600000
TSCA : Yes