accessibility menu, dialog, popup

UserName

CAS RN 304-06-3

OHOHO

CAS RN 304-06-3

IUPAC Name: 2-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 3-phenylsalicylic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: ZJWUEJOPKFYFQD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(C2=CC=CC=C2)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results