missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropylamine 97.0+%, TCI America™

Catalog No. p-7134719
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06645G 5 g
P066425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06645G Supplier TCI America Supplier No. P06645G
Only null left

Chemical Identifiers

CAS 2038-57-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008224
InChI Key LYUQWQRTDLVQGA-UHFFFAOYSA-N
Synonym 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine
PubChem CID 16259
IUPAC Name 3-phenylpropan-1-amine
SMILES NCCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 5 g
UN Number 1760
Formula Weight 135.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Phenylpropylamine
RTECSNumber UI4590000
TSCA No