accessibility menu, dialog, popup

UserName

CAS RN 2038-57-5

IUPAC Name: 3-phenylpropan-1-amine
Synonyms: 3-phenyl propylamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: LYUQWQRTDLVQGA-UHFFFAOYSA-N
SMILES: NCCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)