Learn More
3-Phenylpropylamine, 98+%
CAS: 2038-57-5 | C9H13N | 135.21 g/mol
Supplier: Thermo Scientific Chemicals L0302318
Description
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.
Solubility
Soluble in Chloroform, Ethyl Acetate. Not miscible in water.
Notes
Store away from air and under dry inert gas. Air sensitive. Incompatible with acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
Specifications
| 3-Phenylpropylamine | |
| 0.949 | |
| 90°C (194°F) | |
| C9H13N | |
| MFCD00008224 | |
| UN2735 | |
| Air Sensitive | |
| Soluble in Chloroform,Ethyl Acetate. Not miscible in water. | |
| NCCCC1=CC=CC=C1 | |
| 135.21 | |
| 135.21 |
| 2038-57-5 | |
| 220°C to 221°C | |
| Amine-like | |
| 1.525 | |
| 50 g | |
| 2205526 | |
| 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine | |
| LYUQWQRTDLVQGA-UHFFFAOYSA-N | |
| 3-phenylpropan-1-amine | |
| 16259 | |
| ≥98% |
Chemical Identifiers
| 2038-57-5 | |
| 135.21 | |
| LYUQWQRTDLVQGA-UHFFFAOYSA-N | |
| 16259 | |
| NCCCC1=CC=CC=C1 |
| C9H13N | |
| MFCD00008224 | |
| 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine | |
| 3-phenylpropan-1-amine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 218-012-1
RTECSNumber : UI4590000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only