missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylphenol 97.0+%, TCI America™

Catalog No. p-7134760
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P07685G 5 g
P076825G 25 g
2 options

Catalog No. P07685G

Supplier: TCI America P07685G

Only null left

Chemical Identifiers

CAS 580-51-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002294
InChI Key UBXYXCRCOKCZIT-UHFFFAOYSA-N
Synonym 3-hydroxybiphenyl, 1,1'-biphenyl-3-ol, biphenyl-3-ol, m-phenylphenol, m-hydroxydiphenyl, 3-biphenylol, m-hydroxybiphenyl, 3-hydroxydiphenyl, 3-phenyl phenol, unii-zu11x47h4o
PubChem CID 11381
ChEBI CHEBI:34338
IUPAC Name [1,1'-biphenyl]-3-ol
SMILES OC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 5 g
Formula Weight 170.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenylphenol
EINECSNumber (4)-0019
TSCA Yes