accessibility menu, dialog, popup

UserName

OH

CAS RN 580-51-8

IUPAC Name: [1,1'-biphenyl]-3-ol
Synonyms: m-hydroxydiphenyl 3-hydroxydiphenyl
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: UBXYXCRCOKCZIT-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)