missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylphenol, 90%

Catalog No. p-183438
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC417660050 Supplier Thermo Scientific Chemicals Supplier No. 417660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 580-51-8 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 580-51-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002294
InChI Key UBXYXCRCOKCZIT-UHFFFAOYSA-N
Synonym 3-hydroxybiphenyl, 1,1'-biphenyl-3-ol, biphenyl-3-ol, m-phenylphenol, m-hydroxydiphenyl, 3-biphenylol, m-hydroxybiphenyl, 3-hydroxydiphenyl, 3-phenyl phenol, unii-zu11x47h4o
PubChem CID 11381
ChEBI CHEBI:34338
IUPAC Name 3-phenylphenol
SMILES OC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 72.0°C to 80.0°C
Color Beige-Yellow to White
Boiling Point 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: organic solvents: ethanol,diethyl ether,chlorof,benzene,pyridine
Formula Weight 170.21
Percent Purity 90%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Phenylphenol, 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only